siRNA Details
| Gene Symbol: Pard6g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100973.1 | NP_001094443.1 | 1 | 55 | ||
| GenBank mRNA | EF210570.1 | - | 1 | 55 | ||
| Gene Symbol: LOC100909590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751813.3 | XP_003751861.1 | 238 | |||

Pard6g
Genomic Map