siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107403.1 | NP_001100873.1 | 3 | 2248 | ||
| XM_008771946.1 | XP_008770168.1 | 3 | 2376 | |||
| XM_008771947.1 | XP_008770169.1 | 3 | 2376 | |||
| XM_008771948.1 | XP_008770170.1 | 3 | 2354 | |||
| XM_008771949.1 | XP_008770171.1 | 3 | 2376 | |||

Zfp521
Genomic Map