siRNA Details
| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107441.1 | NP_001100911.1 | 2, 3 | 663 | ||
| XM_006255811.2 | XP_006255873.1 | 3, 4 | 1163 | |||
| GenBank mRNA | FQ211944.1 | - | 2, 3 | 437 | ||
| FQ212467.1 | - | 2, 3 | 436 | |||

Ccsap
Genomic Map