siRNA Details
| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107441.1 | NP_001100911.1 | 1 | 136 | ||
| XM_006255811.2 | XP_006255873.1 | 2 | 636 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccsap| Gene Symbol: Ccsap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107441.1 | NP_001100911.1 | 1 | 136 | ||
| XM_006255811.2 | XP_006255873.1 | 2 | 636 | |||