siRNA Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107492.1 | NP_001100962.1 | 3 | 544 | ||
| XM_006228433.2 | XP_006228495.1 | 2 | 1942 | |||
| XM_006228434.2 | XP_006228496.1 | 2 | 1984 | |||
| XM_006228436.2 | XP_006228498.1 | 2 | 1630 | |||

B3gnt8
Genomic Map