siRNA Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107503.1 | NP_001100973.1 | 4 | 779 | ||
| XM_006228828.2 | XP_006228890.1 | 6 | 760 | |||
| XM_008759152.1 | XP_008757374.1 | 5 | 833 | |||
| XM_008759153.1 | XP_008757375.1 | 5 | 594 | |||
| XM_008759154.1 | XP_008757376.1 | 5 | 594 | |||

Cd22
Genomic Map