siRNA Details
| Gene Symbol: Tshz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107506.1 | NP_001100976.1 | 2 | 2399 | ||
| XM_006228893.1 | XP_006228955.1 | 2 | 2435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tshz3| Gene Symbol: Tshz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107506.1 | NP_001100976.1 | 2 | 2399 | ||
| XM_006228893.1 | XP_006228955.1 | 2 | 2435 | |||