siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107513.1 | NP_001100983.1 | 8 | 1322 | ||
| XM_006229089.2 | XP_006229151.1 | 8 | 1317 | |||
| XM_006229090.2 | XP_006229152.1 | 7 | 1183 | |||
| GenBank mRNA | BC089092.1 | - | 6 | 1122 | ||

Tmem143
Genomic Map