siRNA Details
| Gene Symbol: Pnma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107579.1 | NP_001101049.1 | 1 | 1155 | ||
| XM_008773585.1 | XP_008771807.1 | 4 | 1493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma5| Gene Symbol: Pnma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107579.1 | NP_001101049.1 | 1 | 1155 | ||
| XM_008773585.1 | XP_008771807.1 | 4 | 1493 | |||