siRNA Details
| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227717.2 | XP_006227779.1 | 3 | 497 | ||
| XM_008758652.1 | XP_008756874.1 | 3 | 500 | |||
| XM_008758653.1 | XP_008756875.1 | 3 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc18b1| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227717.2 | XP_006227779.1 | 3 | 497 | ||
| XM_008758652.1 | XP_008756874.1 | 3 | 500 | |||
| XM_008758653.1 | XP_008756875.1 | 3 | 500 | |||