siRNA Details
| Gene Symbol: RGD1560010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107685.1 | NP_001101155.1 | 2 | 482 | ||
| GenBank mRNA | BC162053.1 | - | 2 | 446 | 7113318 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560010| Gene Symbol: RGD1560010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107685.1 | NP_001101155.1 | 2 | 482 | ||
| GenBank mRNA | BC162053.1 | - | 2 | 446 | 7113318 | |