siRNA Details
| Gene Symbol: Fam212b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107713.1 | NP_001101183.1 | 2 | 312 | ||
| XM_006233088.2 | XP_006233150.1 | 3 | 329 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam212b| Gene Symbol: Fam212b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107713.1 | NP_001101183.1 | 2 | 312 | ||
| XM_006233088.2 | XP_006233150.1 | 3 | 329 | |||