siRNA Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002278.1 | NP_001002278.1 | 2 | 238 | ||
| XM_006235290.2 | XP_006235352.2 | 2 | 382 | |||
| GenBank mRNA | AJ781499.1 | - | 2 | 238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pofut1| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002278.1 | NP_001002278.1 | 2 | 238 | ||
| XM_006235290.2 | XP_006235352.2 | 2 | 382 | |||
| GenBank mRNA | AJ781499.1 | - | 2 | 238 | ||