siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017456.1 | NP_001017456.1 | 2 | 322 | ||
| XM_006236201.2 | XP_006236263.1 | 3 | 478 | |||
| GenBank mRNA | BC085351.1 | - | 2 | 322 | 7115168 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc10| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017456.1 | NP_001017456.1 | 2 | 322 | ||
| XM_006236201.2 | XP_006236263.1 | 3 | 478 | |||
| GenBank mRNA | BC085351.1 | - | 2 | 322 | 7115168 | |