siRNA Details
| Gene Symbol: Chchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013431.1 | NP_001013449.1 | 2 | 239 | ||
| GenBank mRNA | BC091407.1 | - | 2 | 239 | 7324948 | |
| FQ220478.1 | - | 2 | 104 | |||
| FQ220795.1 | - | 2 | 134 | |||
| FQ224864.1 | - | 2 | 152 | |||
| FQ229282.1 | - | 2 | 104 | |||
| Gene Symbol: LOC100361898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749083.3 | XP_003749131.1 | 1 | 156 | ||

Chchd4
Genomic Map