siRNA Details
| Gene Symbol: Trnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024261.2 | NP_001019432.1 | 6 | 841 | ||
| XM_008763191.1 | XP_008761413.1 | 6 | 890 | |||
| XM_008763192.1 | XP_008761414.1 | 6 | 846 | |||
| XM_008763193.1 | XP_008761415.1 | 6 | 800 | |||
| GenBank mRNA | BC095841.1 | - | 5 | 766 | 7387807 | |
| BC105612.1 | - | 6 | 841 | 7135310 | ||

Trnt1
Genomic Map