siRNA Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024783.1 | NP_001019954.1 | 9, 10 | 1084 | ||
| XM_006237027.2 | XP_006237089.1 | 8, 9 | 1571 | |||
| XM_006237028.2 | XP_006237090.1 | 8, 9 | 1129 | |||
| GenBank mRNA | BC097951.1 | - | 9, 10 | 1084 | 7442421 | |

Creld1
Genomic Map