siRNA Details
| Gene Symbol: Kdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004268.1 | NP_001004268.1 | 2, 3 | 1111 | ||
| XM_006239015.2 | XP_006239077.1 | 2, 3 | 2152 | |||
| XM_006239016.1 | XP_006239078.1 | 2, 3 | 1336 | |||
| XM_006239017.2 | XP_006239079.1 | 2, 3 | 1949 | |||
| XM_006239018.2 | XP_006239080.1 | 2, 3 | 1242 | |||
| XM_008764133.1 | XP_008762355.1 | 1, 2 | 1578 | |||
| GenBank mRNA | BC079148.1 | - | 2, 3 | 1111 | 7128510 | |

Kdf1
Genomic Map