siRNA Details
| Gene Symbol: Wdtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107908.1 | NP_001101378.1 | 4 | 546 | ||
| XM_008764134.1 | XP_008762356.1 | 4 | 685 | |||
| XM_008764135.1 | XP_008762357.1 | 4 | 342 | |||
| XM_008764136.1 | XP_008762358.1 | 4 | 686 | |||

Wdtc1
Genomic Map