siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 13, 14 | 1228 | ||
| XM_008763918.1 | XP_008762140.1 | 12, 13 | 1143 | |||
| XM_008763919.1 | XP_008762141.1 | 15, 16 | 1255 | |||

Sgip1
Genomic Map