siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763917.1 | XP_008762139.1 | 2 | 408 | ||
| XM_008763918.1 | XP_008762140.1 | 2 | 407 | |||
| XM_008763919.1 | XP_008762141.1 | 2 | 408 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001285862, NM_001285852, NM_001285859, NM_144906, NM_001285860, and others.

Sgip1
Genomic Map