siRNA Details
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763919.1 | XP_008762141.1 | 10, 11 | 934 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_144906, NM_001285852, NM_001285860, NM_001285859, BC017596, and others.

Sgip1
Genomic Map