siRNA Details
| Gene Symbol: Spsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239462.2 | XP_006239524.1 | 3 | 1604 | ||
| XM_008764257.1 | XP_008762479.1 | 3 | 1817 | |||
| XM_008764258.1 | XP_008762480.1 | 2 | 1402 | |||
| XM_008764259.1 | XP_008762481.1 | 3 | 3548 | |||

Spsb1
Genomic Map