siRNA Details
| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134560.1 | NP_001128032.1 | 2, 3 | 700 | ||
| GenBank mRNA | BC158708.1 | - | 2, 3 | 685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rd3l| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134560.1 | NP_001128032.1 | 2, 3 | 700 | ||
| GenBank mRNA | BC158708.1 | - | 2, 3 | 685 | ||