siRNA Details
| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017458.1 | NP_001017458.1 | 5 | 798 | ||
| XM_006241174.2 | XP_006241236.1 | 6 | 860 | |||
| GenBank mRNA | BC092644.1 | - | 5 | 798 | 7318345 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcp11l2| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017458.1 | NP_001017458.1 | 5 | 798 | ||
| XM_006241174.2 | XP_006241236.1 | 6 | 860 | |||
| GenBank mRNA | BC092644.1 | - | 5 | 798 | 7318345 | |