siRNA Details
| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017458.1 | NP_001017458.1 | 6, 7 | 1022 | ||
| XM_006241174.2 | XP_006241236.1 | 7, 8 | 1084 | |||
| GenBank mRNA | BC092644.1 | - | 6, 7 | 1022 | 7318345 | |

Tcp11l2
Genomic Map