siRNA Details
| Gene Symbol: LOC100911483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277222.1 | NP_001264151.1 | 1, 2 | 102 | ||
| GenBank mRNA | FM065403.1 | - | 1, 2 | 98 | ||
| FQ211515.1 | - | 1, 2 | 170 | |||
| FQ223566.1 | - | 1, 2 | 113 | |||
| FQ223617.1 | - | 1, 2 | 112 | |||
| FQ223934.1 | - | 1, 2 | 127 | |||
| FQ224478.1 | - | 1, 2 | 115 | |||

LOC100911483
Genomic Map