siRNA Details
| Gene Symbol: Tymp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012122.1 | NP_001012122.1 | 2 | 196 | ||
| XM_006242209.2 | XP_006242271.1 | 2 | 463 | |||
| XM_008765730.1 | XP_008763952.1 | 2 | 463 | |||
| XM_008765731.1 | XP_008763953.1 | 2 | 463 | |||
| XM_008765732.1 | XP_008763954.1 | 2 | 463 | |||
| XM_008765733.1 | XP_008763955.1 | 2 | 415 | |||
| GenBank mRNA | BC089830.1 | - | 2 | 196 | 7323197 | |

Tymp
Genomic Map