siRNA Details
| Gene Symbol: Olr1307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000519.1 | NP_001000519.1 | 1 | 764 | ||
| GenBank mRNA | X89702.1 | - | 1 | 396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1307| Gene Symbol: Olr1307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000519.1 | NP_001000519.1 | 1 | 764 | ||
| GenBank mRNA | X89702.1 | - | 1 | 396 | ||