siRNA Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007710.1 | NP_001007711.1 | 6 | 1062 | ||
| XM_006243591.2 | XP_006243653.1 | 6 | 1499 | |||
| XM_006243592.2 | XP_006243654.1 | 6 | 2602 | |||
| GenBank mRNA | BC081981.1 | - | 6 | 1062 | 7128571 | |

Pxylp1
Genomic Map