siRNA Details
| Gene Symbol: Cdv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014097.1 | NP_001014119.1 | 2 | 760 | ||
| XM_006243659.2 | XP_006243721.1 | 2 | 666 | |||
| XM_006243660.2 | XP_006243722.1 | 2 | 664 | |||
| XM_008766529.1 | XP_008764751.1 | 2 | 595 | |||
| GenBank mRNA | BC083654.1 | - | 2 | 760 | 7135192 | |

Cdv3
Genomic Map