siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012275.2 | NP_001012275.1 | 3, 4 | 441 | ||
| XM_006243784.2 | XP_006243846.1 | 3, 4 | 909 | |||
| XM_006243785.2 | XP_006243847.1 | 3, 4 | 906 | |||
| XM_008766537.1 | XP_008764759.1 | 3, 4 | 594 | |||
| XM_008766538.1 | XP_008764760.1 | 3, 4 | 597 | |||
| GenBank mRNA | BF284740.1 | - | 3, 4 | 433 | ||

Arih2
Genomic Map