siRNA Details
| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281431.1 | NP_001268360.1 | 5 | 809 | ||
| NM_007196.3 | NP_009127.1 | 6 | 969 | |||
| NM_144505.2 | NP_653088.1 | 6 | 1104 | |||
| NM_144506.2 | NP_653089.1 | 4 | 546 | |||
| NM_144507.2 | NP_653090.1 | 3 | 412 | |||
| NR_104008.1 | 5 | 706 | ||||
| GenBank mRNA | AA320217.1 | - | 2 | 221 | ||
| AB008390.2 | - | 6 | 844 | |||
| AB008927.1 | - | 5 | 946 | |||
| AB009849.1 | - | 6 | 824 | |||
| AF095742.1 | - | 6 | 969 | |||
| AY359036.1 | - | 6 | 955 | |||
| BC040887.1 | - | 7 | 1280 | 5742016 | ||
| BM845575.1 | - | 4 | 512 | |||

KLK8
Genomic Map