siRNA Details
| Gene Symbol: Zfp105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012128.2 | NP_001012128.1 | 4 | 614 | ||
| GenBank mRNA | BC078921.1 | - | 3 | 413 | ||
| BC088185.1 | - | 3 | 450 | 7367875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp105| Gene Symbol: Zfp105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012128.2 | NP_001012128.1 | 4 | 614 | ||
| GenBank mRNA | BC078921.1 | - | 3 | 413 | ||
| BC088185.1 | - | 3 | 450 | 7367875 | ||