siRNA Details
| Gene Symbol: Sult1c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013177.2 | NP_001013195.2 | 994 | |||
| GenBank mRNA | BC092564.1 | - | 994 | 7372924 | ||
| FQ209913.1 | - | 988 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c2a| Gene Symbol: Sult1c2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013177.2 | NP_001013195.2 | 994 | |||
| GenBank mRNA | BC092564.1 | - | 994 | 7372924 | ||
| FQ209913.1 | - | 988 | ||||