siRNA Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108215.1 | NP_001101685.1 | 4, 5 | 459 | ||
| XM_006244823.2 | XP_006244885.1 | 4, 5 | 451 | |||
| XM_006244824.2 | XP_006244886.1 | 4, 5 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd9| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108215.1 | NP_001101685.1 | 4, 5 | 459 | ||
| XM_006244823.2 | XP_006244885.1 | 4, 5 | 451 | |||
| XM_006244824.2 | XP_006244886.1 | 4, 5 | 595 | |||