siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB610197.1 | - | 3 | 415 | ||
| Gene Symbol: Ndufa10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182671.2 | NP_872612.1 | 1 | 377 | ||
| GenBank mRNA | AY260163.1 | - | 1 | 377 | ||

Ndufa10
Genomic Map