siRNA Details
| Gene Symbol: Myod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176079.1 | NP_788268.1 | 3 | 1681 | ||
| GenBank mRNA | BC127480.1 | - | 3 | 1662 | 8366406 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myod1| Gene Symbol: Myod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176079.1 | NP_788268.1 | 3 | 1681 | ||
| GenBank mRNA | BC127480.1 | - | 3 | 1662 | 8366406 | |