siRNA Details
| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003706.3 | NP_001003706.2 | 3 | 303 | ||
| GenBank mRNA | AB101676.1 | - | 3 | 319 | ||
| BG661061.1 | - | 3 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc21| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003706.3 | NP_001003706.2 | 3 | 303 | ||
| GenBank mRNA | AB101676.1 | - | 3 | 319 | ||
| BG661061.1 | - | 3 | 303 | |||