siRNA Details
| Gene Symbol: RGD1311343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108270.1 | NP_001101740.1 | 1 | 260 | ||
| XM_006246018.2 | XP_006246080.1 | 1 | 304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311343| Gene Symbol: RGD1311343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108270.1 | NP_001101740.1 | 1 | 260 | ||
| XM_006246018.2 | XP_006246080.1 | 1 | 304 | |||