siRNA Details
| Gene Symbol: Sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_340803.7 | XP_340804.4 | 1 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sap30l| Gene Symbol: Sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_340803.7 | XP_340804.4 | 1 | 282 | ||