siRNA Details
| Gene Symbol: Lztr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108326.2 | NP_001101796.1 | 19 | 2323 | ||
| XM_006248707.2 | XP_006248769.1 | 19 | 2636 | |||
| GenBank mRNA | BC169071.1 | - | 19 | 2323 | 7938530 | |
| FQ211175.1 | - | 2 | 214 | |||
| FQ211947.1 | - | 18 | 3036 | |||
| FQ218020.1 | - | 4 | 442 | |||

Lztr1
Genomic Map