siRNA Details
| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025125.2 | NP_001020296.2 | 5 | 442 | ||
| GenBank mRNA | BC091176.1 | - | 5 | 448 | 7322044 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sumf2| Gene Symbol: Sumf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025125.2 | NP_001020296.2 | 5 | 442 | ||
| GenBank mRNA | BC091176.1 | - | 5 | 448 | 7322044 | |