siRNA Details
| Gene Symbol: Slc25a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013187.1 | NP_001013205.1 | 6 | 562 | ||
| XM_008770873.1 | XP_008769095.1 | 6 | 562 | |||
| GenBank mRNA | AY724532.1 | - | 6 | 534 | ||
| BC087106.1 | - | 6 | 562 | 7193157 | ||

Slc25a30
Genomic Map