siRNA Details
| Gene Symbol: Zic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108391.1 | NP_001101861.1 | 3 | 1149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zic5| Gene Symbol: Zic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108391.1 | NP_001101861.1 | 3 | 1149 | ||