siRNA Details
| Gene Symbol: Cd83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108410.1 | NP_001101880.1 | 5 | 587 | ||
| GenBank mRNA | BC166582.1 | - | 5 | 498 | 7385133 | |
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Cd83
Cd83| Gene Symbol: Cd83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108410.1 | NP_001101880.1 | 5 | 587 | ||
| GenBank mRNA | BC166582.1 | - | 5 | 498 | 7385133 | |
 Back To Top
 Back To Top