siRNA Details
| Gene Symbol: LOC688637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254612.2 | XP_006254674.1 | 23 | 2952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688637| Gene Symbol: LOC688637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254612.2 | XP_006254674.1 | 23 | 2952 | ||