siRNA Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012167.1 | NP_001012167.1 | 10, 11 | 1264 | ||
| XM_006228578.2 | XP_006228640.1 | 12, 13 | 1593 | |||
| XM_006228579.2 | XP_006228641.1 | 12, 13 | 1541 | |||
| XM_006228580.2 | XP_006228642.1 | 11, 12 | 1699 | |||
| XM_006228581.2 | XP_006228643.1 | 12, 13 | 1571 | |||
| XM_008759165.1 | XP_008757387.1 | 11, 12 | 1484 | |||
| GenBank mRNA | BC089987.1 | - | 10, 11 | 1264 | 7318612 | |

Pld3
Genomic Map