siRNA Details
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751958.3 | XP_003752006.2 | 6 | 1046 | ||
XM_006256396.2 | XP_006256458.1 | 6 | 1091 |
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108532.1 | NP_001102002.1 | 697 | |||
XM_006256376.2 | XP_006256438.1 | 7 | 2352 | |||
XM_006256377.2 | XP_006256439.1 | 7 | 2291 | |||
XM_006256378.2 | XP_006256440.1 | 7 | 2271 | |||
XM_006256379.2 | XP_006256441.1 | 7 | 2931 | |||
XM_006256380.2 | XP_006256442.1 | 6 | 1039 | |||
GenBank mRNA | FQ215209.1 | - | 7 | 921 |
