siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191847.1 | NP_001178776.1 | 15 | 1372 | ||
| XM_008761056.1 | XP_008759278.1 | 15 | 1374 | |||
| GenBank mRNA | FQ211604.1 | - | 1389 | |||
| FQ217493.1 | - | 1357 | ||||

Mfsd1
Genomic Map